accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34334-04-8

IUPAC Name: 4-[2-(morpholin-4-yl)ethoxy]benzonitrile
Synonyms: 4-[2-(morpholin-4-yl)ethoxy]benzonitrile
Molecular Weight (g/mol): 232.28
Molecular Formula: C13H16N2O2
InChi Key: KQTJRPZKCNRDQC-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(OCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results