accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClFF

CAS RN 344-07-0

IUPAC Name: 1-chloro-2,3,4,5,6-pentafluorobenzene
Synonyms: chloropentafluorobenzene
Molecular Weight (g/mol): 202.51
Molecular Formula: C6ClF5
InChi Key: KGCDGLXSBHJAHZ-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(Cl)C(F)=C1F
Boiling Point 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)