Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloropentafluorobenzene, 98+%

Catalog No. AAA1538430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538430 250 g
AAA1538409 10 g
AAA1538418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1538430 Supplier Thermo Scientific Chemicals Supplier No. A1538430
  • $613.00
    $521.65 / Each of 1

    Save $91.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 344-07-0 | C6ClF5 | 202.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-07-0
Molecular Formula C6ClF5
Molecular Weight (g/mol) 202.51
MDL Number MFCD00000534
InChI Key KGCDGLXSBHJAHZ-UHFFFAOYSA-N
Synonym chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
PubChem CID 9579
IUPAC Name 1-chloro-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Cl)C(F)=C1F

Specifications

Density 1.568
Boiling Point 116°C to 118°C
Refractive Index 1.424
Quantity 250 g
Beilstein 1819389
Formula Weight 202.51
Percent Purity ≥98%
Chemical Name or Material Chloropentafluorobenzene
Hazard Category H331
Precautionary Statement P261-P271-P304+P340-P311-P501c
EINECSNumber 206-450-6
RTECSNumber CZ1225500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.