accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 344-20-7

IUPAC Name: 2,6-dibromo-4-fluorophenol
Synonyms: 2,6-dibromo-4-fluorophenol
Molecular Weight (g/mol): 269.9
Molecular Formula: C6H3Br2FO
InChi Key: RRAZCUUOWIDAJS-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(F)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results