Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-4-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. D35315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35315G 5 g
D353125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35315G Supplier TCI America Supplier No. D35315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 344-20-7
Molecular Formula C6H3Br2FO
Molecular Weight (g/mol) 269.895
MDL Number MFCD00002317
InChI Key RRAZCUUOWIDAJS-UHFFFAOYSA-N
Synonym phenol, 2,6-dibromo-4-fluoro, 2,6-dibromo-4-fluoro-phenol, pubchem2868, acmc-209i7j, ksc494q8h, emolecules 492569, 1-bromo-3,4,5-trifluorobezene, attercop-chm at113643, 2,6-dibromo-4-fluorophenol
PubChem CID 67654
IUPAC Name 2,6-dibromo-4-fluorophenol
SMILES C1=C(C=C(C(=C1Br)O)Br)F

Specifications

Melting Point 56°C
Color Yellow
Quantity 5 g
Formula Weight 269.90
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-4-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.