accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34403-52-6

H3CNCH3NH3

CAS RN 34403-52-6

IUPAC Name: [4-(dimethylamino)phenyl]methanaminium
Synonyms: [4-(dimethylamino)phenyl]methanaminium
Molecular Weight (g/mol): 151.23
Molecular Formula: C9H15N2
InChi Key: PDJZOFLRRJQYBF-UHFFFAOYSA-O
SMILES: CN(C)C1=CC=C(C[NH3+])C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)