accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOCH

CAS RN 34450-18-5

IUPAC Name: octadec-17-ynoic acid
Synonyms: 17-octadecynoic acid
Molecular Weight (g/mol): 280.45
Molecular Formula: C18H32O2
InChi Key: DZIILFGADWDKMF-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCCCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results