Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17-ODYA, Tocris Bioscience™
SDP

Catalog No. 060710
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-071-0 Supplier R&D Systems Supplier No. 0607/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LTB ω-hydroxylase inhibitor

Potent suicide inhibitor of LTB4 ω-hydroxylase.

Chemical Identifiers

CAS 34450-18-5
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
InChI Key DZIILFGADWDKMF-UHFFFAOYSA-N
Synonym 17-octadecynoic acid, 17-odya, alkynyl stearic acid, tocris-0607, bml3-c04, 17-odya 17-octadecynoic acid, 17-octadecynoic acid gc
PubChem CID 1449
IUPAC Name octadec-17-ynoic acid
SMILES C#CCCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 10 mg
Formula Weight 280.45
Percent Purity >98%
Chemical Name or Material 17-ODYA
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.