accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CNS

CAS RN 344790-87-0

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; cyanosulfanide
Synonyms: 1-butyl-3-methylimidazolium thiocyanate
Molecular Weight (g/mol): 197.3
Molecular Formula: C9H15N3S
InChi Key: SIXHYMZEOJSYQH-UHFFFAOYSA-M
SMILES: [S-]C#N.CCCCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results