Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Thiocyanate 97.0+%, TCI America™
SDP

Catalog No. B40915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40915G 5 g
B409125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40915G Supplier TCI America Supplier No. B40915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 344790-87-0
Molecular Formula C9H15N3S
Molecular Weight (g/mol) 197.3
MDL Number MFCD06798181
InChI Key SIXHYMZEOJSYQH-UHFFFAOYSA-M
PubChem CID 11424151
IUPAC Name 1-butyl-3-methylimidazol-3-ium;thiocyanate
SMILES CCCCN1C=C[N+](=C1)C.C(#N)[S-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 197.30
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.