accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2FFF

CAS RN 34486-24-3

IUPAC Name: 6-(trifluoromethyl)pyridin-2-amine
Synonyms: 6-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: NFYYDQMFURPHCC-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=N1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)