Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-(trifluoromethyl)pyridine, 98+%

Catalog No. AAL19443ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19443ME 500 mg
AAL19443MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19443ME Supplier Thermo Scientific Chemicals Supplier No. L19443ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 34486-24-3 | C6H5F3N2 | 162.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34486-24-3
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD03093868
InChI Key NFYYDQMFURPHCC-UHFFFAOYSA-N
Synonym 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w
PubChem CID 821024
IUPAC Name 6-(trifluoromethyl)pyridin-2-amine
SMILES C1=CC(=NC(=C1)N)C(F)(F)F

Specifications

Melting Point 86°C to 89°C
Quantity 500 mg
UN Number UN2811
Beilstein 473259
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 2-Amino-6-(trifluoromethyl)pyridine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.