accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34491-29-7

IUPAC Name: 2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid
Synonyms: 2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 243.71
Molecular Formula: C10H10ClNO2S
InChi Key: LESQASCTNMKKPZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CSC(N1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results