accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 345-17-5

IUPAC Name: 1-chloro-4-fluoro-2-nitrobenzene
Synonyms: 1-chloro-4-fluoro-2-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: DVXDJQKEEKXJBW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results