accessibility menu, dialog, popup

UserName

Viewing profile as user:
OF

CAS RN 345-69-7

IUPAC Name: (3-fluorophenyl)(phenyl)methanone
Synonyms: (3-fluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 200.21
Molecular Formula: C13H9FO
InChi Key: NCIYZALOQBXNLW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results