accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3CH3H3C

CAS RN 3452-97-9

IUPAC Name: 3,5,5-trimethylhexan-1-ol
Synonyms: trimethylhexanol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: BODRLKRKPXBDBN-UHFFFAOYNA-N
SMILES: CC(CCO)CC(C)(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)