Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5,5-Trimethyl-1-hexanol

Catalog No. 28948525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-948-525ML 25 mL
28-948-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-948-525ML Supplier Sigma Aldrich Supplier No. 28948525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A dermatologic and toxicologic analysis of 3,5,5-trimethyl-1-hexanol (when used as a fragrance ingredient) was reported.

Specifications

CAS 3452-97-9
Density 0.824 g/mL (at 25°C (literature))
Boiling Point 193°C to 194°C (lit.)
Molecular Formula C9H20O
Refractive Index n20/D 1.432 (literature)
Linear Formula (CH3)3 CCH2CH(CH3)CH2CH2OH
MDL Number MFCD00036138
Quantity 25 mL
Molecular Weight (g/mol) 144.25
Percent Purity 85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.