accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34552-13-1

CH3NClH2N

CAS RN 34552-13-1

IUPAC Name: 2-chloro-5-methylpyridin-3-amine
Synonyms: 2-chloro-5-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: CQKIBEOVARIBDN-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results