accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3NN(E/Z)

CAS RN 3457-98-5

IUPAC Name: 4-(2-phenyldiazen-1-yl)anilinium chloride
Synonyms: 4-(2-phenyldiazen-1-yl)anilinium chloride
Molecular Weight (g/mol): 233.7
Molecular Formula: C12H12ClN3
InChi Key: WMNTYZIRLUBHEE-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results