Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoazobenzene Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. A077125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A077125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A077125G Supplier TCI America Supplier No. A077125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3457-98-5
Molecular Formula C12H12ClN3
Molecular Weight (g/mol) 233.699
MDL Number MFCD00012994
InChI Key WMNTYZIRLUBHEE-UHFFFAOYSA-N
Synonym 4-aminoazobenzene hydrochloride, 4-phenyldiazenyl aniline hydrochloride, benzenamine, 4-phenylazo-, monohydrochloride, 4-phenylazo aniline hydrochloride, benzenamine, 4-2-phenyldiazenyl-, hydrochloride 1:1, p-aminoazobenzene hcl, acmc-1aik2, p-aminoazobenzolhydrochlorid, p-aminoazobenzene hcl, pract, benzenamine,4-2-phenyldiazenyl-, hydrochloride 1:1
PubChem CID 12203296
IUPAC Name 4-phenyldiazenylaniline;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N.Cl

Specifications

Melting Point 213°C
Color Blue-Purple
Quantity 25 g
Formula Weight 233.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoazobenzene Hydrochloride
RTECSNumber BY8235000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.