accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34582-32-6

IUPAC Name: 4-(tert-butoxy)-3-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(tert-butoxy)-3-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
Molecular Weight (g/mol): 289.33
Molecular Formula: C13H23NO6
InChi Key: RAUQRYTYJIYLTF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)