Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate 95.0+%, TCI America™
SDP

Catalog No. B46185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46185G 5 g
B46181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46185G Supplier TCI America Supplier No. B46185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34582-32-6
Molecular Formula C13H23NO6
Molecular Weight (g/mol) 289.328
MDL Number MFCD00038272
InChI Key RAUQRYTYJIYLTF-QMMMGPOBSA-N
Synonym boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu
PubChem CID 7010517
IUPAC Name (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C

Specifications

Melting Point 103°C
Color White
Quantity 5 g
Formula Weight 289.33
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.