accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)ONH2OHHSHCl

CAS RN 345909-32-2

IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
Synonyms: cysteine hcl
Molecular Weight (g/mol): 157.61
Molecular Formula: C3H8ClNO2S
InChi Key: IFQSXNOEEPCSLW-QYLAHFPANA-N
SMILES: Cl.N[C@@H](CS)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results