accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 345968-40-3

H3COOCH3

CAS RN 345968-40-3

IUPAC Name: 1-ethoxy-4-(hexyloxy)benzene
Synonyms: 1-ethoxy-4-(hexyloxy)benzene
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: MWIZWOMGGAKSLB-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(OCC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results