Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethoxy-4-hexyloxybenzene, TCI America™
SDP

Catalog No. E05205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05205G Supplier TCI America Supplier No. E05205G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 345968-40-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD02093468
InChI Key MWIZWOMGGAKSLB-UHFFFAOYSA-N
PubChem CID 44630283
IUPAC Name 1-ethoxy-4-hexoxybenzene
SMILES CCCCCCOC1=CC=C(C=C1)OCC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 222.33
Physical Form Crystals
Chemical Name or Material 1-Ethoxy-4-hexyloxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.