accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34598-33-9

IUPAC Name: 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
Synonyms: 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
Molecular Weight (g/mol): 492.13
Molecular Formula: C11H5F17O2
InChi Key: JZRCRCFPVAXHHQ-UHFFFAOYSA-N
SMILES: OC(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results