Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2H,2H,3H,3H-Heptadecafluoroundecanoic Acid 97.0+%, TCI America™
SDP

Catalog No. H15021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15021G Supplier TCI America Supplier No. H15021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34598-33-9
Molecular Formula C11H5F17O2
Molecular Weight (g/mol) 492.132
MDL Number MFCD04038353
InChI Key JZRCRCFPVAXHHQ-UHFFFAOYSA-N
Synonym 2H,2H,3H,3H-Perfluoroundecanoic Acid, 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic Acid
PubChem CID 2782413
IUPAC Name 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
SMILES C(CC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(=O)O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 492.13
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2H,2H,3H,3H-Heptadecafluoroundecanoic Acid
EINECSNumber (2)-1182
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.