accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOH

CAS RN 34637-22-4

IUPAC Name: benzyl N-(3-hydroxypropyl)carbamate
Synonyms: benzyl N-(3-hydroxypropyl)carbamate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: WXQCFKYWSKKNKY-UHFFFAOYSA-N
SMILES: OCCCNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results