accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SPSOCH3OClCl

CAS RN 34643-46-4

IUPAC Name: O-2,4-dichlorophenyl O-ethyl (propylsulfanyl)phosphonothioate
Synonyms: prothiofos epa pesticide chemical code 128858 dichlorpropaphos bideron toyothion prothiophos toyodan tukuthion caswell no. 714H tokuthion
Molecular Weight (g/mol): 345.23
Molecular Formula: C11H15Cl2O2PS2
InChi Key: FITIWKDOCAUBQD-UHFFFAOYNA-N
SMILES: CCCSP(=S)(OCC)OC1=CC=C(Cl)C=C1Cl
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)