accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3F

CAS RN 346656-39-1

IUPAC Name: 2-(2-fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-(2-fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 208.04
Molecular Formula: C11H14BFO2
InChi Key: ZNFBLQHZFSLYQB-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=CC=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results