Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane 98.0+%, TCI America™
SDP

Catalog No. F05311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05311G Supplier TCI America Supplier No. F05311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 346656-39-1
Molecular Formula C11H14BFO2
Molecular Weight (g/mol) 208.039
InChI Key ZNFBLQHZFSLYQB-UHFFFAOYSA-N
Synonym 1-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-2-fluorobenzene, 2-Fluorophenylboronic Acid Neopentyl Glycol Ester
PubChem CID 10398127
IUPAC Name 2-(2-fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=CC=C2F

Specifications

Melting Point 40°C
Quantity 1 g
Formula Weight 208.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.