accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 34671-83-5

IUPAC Name: 2,2'-bipyrimidine
Synonyms: 2,2'-bipyrimidine
Molecular Weight (g/mol): 158.16
Molecular Formula: C8H6N4
InChi Key: HKOAFLAGUQUJQG-UHFFFAOYSA-N
SMILES: C1=CN=C(N=C1)C1=NC=CC=N1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)