accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO

CAS RN 3470-98-2

IUPAC Name: 1-butylpyrrolidin-2-one
Synonyms: 2-pyrrolidinone, 1-butyl-
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: BNXZHVUCNYMNOS-UHFFFAOYSA-N
SMILES: CCCCN1CCCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results