Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. B12545ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12545ML 5 mL
B125425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B12545ML Supplier TCI America Supplier No. B12545ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3470-98-2
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00020876
InChI Key BNXZHVUCNYMNOS-UHFFFAOYSA-N
Synonym N-Butyl-2-azacyclopentanone
PubChem CID 18984
IUPAC Name 1-butylpyrrolidin-2-one
SMILES CCCCN1CCCC1=O

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 mL
Formula Weight 141.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-2-pyrrolidone
RTECSNumber UY5746000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.