accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34702-59-5

IUPAC Name: (2S)-2-amino-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Molecular Weight (g/mol): 255.14
Molecular Formula: C9H6F5NO2
InChi Key: YYTDJPUFAVPHQA-VKHMYHEASA-N
SMILES: N[C@@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results