Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoro-L-phenylalanine 95.0+%, TCI America™
SDP

Catalog No. P2085200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2085200MG 200 mg
P20851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2085200MG Supplier TCI America Supplier No. P2085200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34702-59-5
Molecular Formula C9H6F5NO2
Molecular Weight (g/mol) 255.14
MDL Number MFCD01860881
InChI Key YYTDJPUFAVPHQA-VKHMYHEASA-N
PubChem CID 2761496
IUPAC Name (2S)-2-amino-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
SMILES N[C@@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O

Specifications

Melting Point 258°C
Color White-Yellow
Quantity 200 mg
Formula Weight 255.14
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluoro-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.