accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34737-89-8

IUPAC Name: (3S)-1-benzyl-3-methyl-4-oxopiperidin-1-ium
Synonyms: (3S)-1-benzyl-3-methyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol): 204.29
Molecular Formula: C13H18NO
InChi Key: OVQAJYCAXPHYNV-NSHDSACASA-O
SMILES: C[C@H]1C[NH+](CC2=CC=CC=C2)CCC1=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)