Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-methyl-4-piperidone, 99%

Catalog No. AC134040250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC134040250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC134040250 Supplier Thermo Scientific Chemicals Supplier No. 134040250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 34737-89-8 | C13H17NO | 203.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34737-89-8
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.28
MDL Number MFCD00044806
InChI Key OVQAJYCAXPHYNV-UHFFFAOYSA-N
Synonym 1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
PubChem CID 560965
IUPAC Name 1-benzyl-3-methylpiperidin-4-one
SMILES CC1CN(CCC1=O)CC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5315 to 1.5335
Quantity 25 g
Beilstein 21,V,6,483
Formula Weight 203.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-methyl-4-piperidone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 252-176-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.