accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34784-04-8

IUPAC Name: 5-bromoisoquinoline
Synonyms: 5-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: CYJZJGYYTFQQBY-UHFFFAOYSA-N
SMILES: BrC1=C2C=CN=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results