Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoisoquinoline 96.0+%, TCI America™
SDP

Catalog No. B33135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33135G 5 g
B33131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33135G Supplier TCI America Supplier No. B33135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34784-04-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD01646405
InChI Key CYJZJGYYTFQQBY-UHFFFAOYSA-N
Synonym isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg
PubChem CID 736487
IUPAC Name 5-bromoisoquinoline
SMILES BrC1=C2C=CN=CC2=CC=C1

Specifications

Melting Point 82°C
Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 208.06
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.