accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHF

CAS RN 347867-75-8

IUPAC Name: 5-(4-fluorophenoxy)pentanoic acid
Synonyms: 5-(4-fluorophenoxy)pentanoic acid
Molecular Weight (g/mol): 212.22
Molecular Formula: C11H13FO3
InChi Key: SNUHBWJBUYDESY-UHFFFAOYSA-N
SMILES: OC(=O)CCCCOC1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results