Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Fluorophenoxy)valeric Acid 98.0+%, TCI America™
SDP

Catalog No. F04105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04105G Supplier TCI America Supplier No. F04105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 347867-75-8
Molecular Formula C11H13FO3
Molecular Weight (g/mol) 212.22
MDL Number MFCD02093469
InChI Key SNUHBWJBUYDESY-UHFFFAOYSA-N
PubChem CID 22556441
IUPAC Name 5-(4-fluorophenoxy)pentanoic acid
SMILES C1=CC(=CC=C1OCCCCC(=O)O)F

Specifications

Melting Point 80°C
Color Red-White
Quantity 5 g
Formula Weight 212.22
Percent Purity ≥98.0% (T,GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Fluorophenoxy)valeric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.