accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOONHOO

CAS RN 3479-47-8

IUPAC Name: 4-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol): 357.36
Molecular Formula: C19H19NO6
InChi Key: VUKCNAATVIWRTF-UHFFFAOYNA-N
SMILES: OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)