Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester, 98+%, Thermo Scientific™

Catalog No. AAL0843606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0843606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0843606 Supplier Thermo Scientific Chemicals Supplier No. L0843606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used as a pharmaceutical intermediate and in food industry.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3479-47-8
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00037820
InChI Key VUKCNAATVIWRTF-UHFFFAOYNA-N
Synonym z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
PubChem CID 47602
IUPAC Name (2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 105°C to 107°C
Quantity 5 g
Beilstein 2065292
Solubility Information Insoluble in water.
Formula Weight 357.37
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester
EINECSNumber 222-456-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.