accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOOFFF

CAS RN 348-75-4

IUPAC Name: 4,4,4-trifluoro-3-methylbutanoic acid
Synonyms: 4,4,4-trifluoro-3-methylbutanoic acid
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H7F3O2
InChi Key: FFZMMBKGTNDVRX-UHFFFAOYNA-N
SMILES: CC(CC(O)=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)