Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)butyric acid, 97%

Catalog No. AAL1216006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1216006 5 g
AAL1216003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1216006 Supplier Thermo Scientific Chemicals Supplier No. L1216006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 348-75-4 | C5H7F3O2 | 156.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-75-4
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD00039533
InChI Key FFZMMBKGTNDVRX-UHFFFAOYSA-N
Synonym 3-trifluoromethylbutyric acid, 3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-methyl-butanoic acid, 3-trifluoromethyl butanoic acid, 3-methyl-4,4,4-trifluorobutyric acid, butanoic acid, 4,4,4-trifluoro-3-methyl, zlchem 1052, acmc-1aijc, 3-trifluoromethylbutanoic acid
PubChem CID 136174
IUPAC Name 4,4,4-trifluoro-3-methylbutanoic acid
SMILES CC(CC(=O)O)C(F)(F)F

Specifications

Boiling Point 90°C to 91°C (25 mmHg)
Odor Pungent
Refractive Index 1.359
Quantity 5 g
UN Number UN3265
Beilstein 1760574
Formula Weight 156.11
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)butyric acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.