accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34801-14-4

IUPAC Name: 4-(5-bromothiophen-2-yl)-1,3-thiazol-2-amine
Synonyms: 4-(5-bromothiophen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 261.16
Molecular Formula: C7H5BrN2S2
InChi Key: MMZHPDCFNLBMBY-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results