Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(5-Bromo-2-thienyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Catalog No. BTB09097DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09097DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09097DA Supplier Thermo Scientific Chemicals Supplier No. BTB09097DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34801-14-4
Molecular Formula C7H5BrN2S2
Molecular Weight (g/mol) 261.155
MDL Number MFCD01812622
InChI Key MMZHPDCFNLBMBY-UHFFFAOYSA-N
Synonym 4-5-bromothiophen-2-yl thiazol-2-amine, 4-5-bromothiophen-2-yl-1,3-thiazol-2-amine, 4-5-bromothiophen-2-yl thiazol-2-ylamine, 4-5-bromo-2-thienyl-1,3-thiazol-2-amine, 4-5-bromo-thiophen-2-yl-thiazol-2-ylamine, 2-amino-4-5-bromothien-2-yl thiazole, 4-5-bromothien-2-yl-1,3-thiazol-2-amine, 4-5-bromo-2-thienyl-1,3-thiazole-2-ylamine, 2-thiazolamine, 4-5-bromo-2-thienyl, maybridge1_001930
PubChem CID 725239
IUPAC Name 4-(5-bromothiophen-2-yl)-1,3-thiazol-2-amine
SMILES C1=C(SC(=C1)Br)C2=CSC(=N2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 261.17
Percent Purity 97%
Chemical Name or Material 4-(5-Bromo-2-thienyl)-1, 3-thiazol-2-amine
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.