accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34805-21-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methanesulfinylbutanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-4-methanesulfinylbutanoic acid|BOC-MET(O)-OH
Molecular Weight (g/mol): 265.32
Molecular Formula: C10H19NO5S
InChi Key: FVSDTYGQCVACMH-ISJKBYAMSA-N
SMILES: CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)