accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OOSOOOOSOOOZr

CAS RN 34806-73-0

IUPAC Name: zirconium(4+) hydrate disulfate
Synonyms: zirconium(4+) hydrate disulfate
Molecular Weight (g/mol): 301.35
Molecular Formula: H2O9S2Zr
InChi Key: UXRMCZYAVOHQNB-UHFFFAOYSA-J
SMILES: O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)